Learn More
Axygen™ Conical-Bottom Screw-Cap Tubes, Attached Clear Looped Caps
Graduated tubes with conical bottom
Brand: Axygen™ SCT-150-LP-C
Code : A0
Additional Details : Weight : 7.30000kg
Description
- Writing area for sample identification
- Caps close with one hand
Specifications
20,000 x g | |
No | |
Clear | |
For sample identification | |
Non-sterile |
Screw Cap | |
1.5 mL | |
Polypropylene | |
Attached clear looped cap | |
Conical |