missing translation for 'onlineSavingsMsg'
Learn More

ATAGO™ Master-2M Refractometer

The MASTER-M series is a good alternative choice when the water resistant feature is not a priority and the Temperature Compensation will be carried out manually. The ATAGO™ Master-2M Refractometer is a Mid-range model of the MASTER-M, and a successor model to the conventional N-2E.

Brand:  ATAGO™ 2323

Code : A0

Additional Details : Weight : 0.80000kg

Product Code. 10314513

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »


Description

Description

  • Measurement range Brix 28.0 to 62.0%
  • Constructed of metal; for salty and acidic samples, choose the plastic body (MASTER-2PM)
  • Not equipped with the Automatic Temperature Compensation feature
Specifications

Specifications

Refractometer
Master-2M
Min. Scale ±0.2%
28.0 to 62.0 Brix
±0.2%
Brix
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers