missing translation for 'onlineSavingsMsg'
Learn More

Arsenic Standard For ICP, 1000μg/mL in 2% HNO3, Assurance Grade, SPEX CertiPrep™

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

£89.45 - £160.00

Chemical Identifiers

CAS 7440-38-2, 7697-37-2, 7732-18-5
Molecular Formula As
Molecular Weight (g/mol) 74.92
MDL Number MFCD00085309
InChI Key RQNWIZPPADIBDY-UHFFFAOYSA-N
PubChem CID 5359596
ChEBI CHEBI:27563
IUPAC Name arsenic
SMILES [As]
View More Specs

Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10171762
View Documents
SPEX Certiprep
PLAS2-2Y
125 mL Triple-leached LDPE bottle
£89.45
125mL
Please to purchase this item. Need a web account? Register with us today!
11468619
View Documents
SPEX Certiprep
PLAS2-2T
250 mL Triple-leached LDPE bottle
£106.00
250mL
Please to purchase this item. Need a web account? Register with us today!
10224242
View Documents
SPEX Certiprep
PLAS2-2X
500 mL Triple-leached LDPE bottle
£160.00
500mL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

AnalyteConcentration
As1000μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Specifications

Chemical Identifiers

7440-38-2, 7697-37-2, 7732-18-5
74.92
RQNWIZPPADIBDY-UHFFFAOYSA-N
CHEBI:27563
[As]
As
MFCD00085309
5359596
arsenic

Specifications

7440-38-2, 7697-37-2, 7732-18-5
Certified Reference Material
As
MFCD00085309
RQNWIZPPADIBDY-UHFFFAOYSA-N
arsenic
5359596
Assurance
Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
2% HNO3
AA, ICP
[As]
74.92
CHEBI:27563
Arsenic Standard
Videos
Safety and Handling

Safety and Handling

Causes severe skin burns and eye damage.

missing translation for 'dotInformation' : Hazardous

Recommended Storage : Ambient

Product Identifier
  • 1000

Signal Word
  • Danger||Warning

Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.

Precautionary Statement
  • P101-If medical advice is needed, have product container or label at hand.
  • P102-Keep out of reach of children.
  • P103-Read label before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P264-Wash thoroughly after handling.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P321-Specific treatment (see on this label).
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

Supplemental information
  • MIXTURE LIST-Contains : nitric acid, arsenic

SDS
Documents

Documents

Product Certifications

Product intended for laboratory use only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.