missing translation for 'onlineSavingsMsg'
Learn More

Arecoline hydrobromide, 98+%, Thermo Scientific Chemicals

£25.40 - £289.44

Chemical Identifiers

CAS 300-08-3
Molecular Formula C8H14BrNO2
Molecular Weight (g/mol) 236.11
MDL Number MFCD00039041
InChI Key AXOJRQLKMVSHHZ-UHFFFAOYSA-N
Synonym arecoline hydrobromide, arecoline bromide, taeniolin, arecoline hbr, methyl 1-methyl-1,2,5,6-tetrahydropyridine-3-carboxylate hydrobromide, arekolinhydrobromid german, unii-24s79b9cx7, methyl 1,2,5,6-tetrahydro-1-methylnicotinate hydrobromide, n-methyltetrahydronicotinic acid methyl ester hydrobromide
PubChem CID 9301
IUPAC Name methyl 1-methyl-3,6-dihydro-2H-pyridine-5-carboxylate;hydrobromide
SMILES [H+].[Br-].COC(=O)C1=CCCN(C)C1
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10100032
View Documents
Thermo Scientific Acros
250130050
5 g Glass bottle
£25.40
5g
Please to purchase this item. Need a web account? Register with us today!
10297250
View Documents
Thermo Scientific Acros
250130250
25 g Glass bottle
£99.50
25g
Please to purchase this item. Need a web account? Register with us today!
10023813
View Documents
Thermo Scientific Acros
250131000
100 g Glass bottle
£289.44
100g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

300-08-3
236.11
AXOJRQLKMVSHHZ-UHFFFAOYSA-N
9301
[H+].[Br-].COC(=O)C1=CCCN(C)C1
C8H14BrNO2
MFCD00039041
arecoline hydrobromide, arecoline bromide, taeniolin, arecoline hbr, methyl 1-methyl-1,2,5,6-tetrahydropyridine-3-carboxylate hydrobromide, arekolinhydrobromid german, unii-24s79b9cx7, methyl 1,2,5,6-tetrahydro-1-methylnicotinate hydrobromide, n-methyltetrahydronicotinic acid methyl ester hydrobromide
methyl 1-methyl-3,6-dihydro-2H-pyridine-5-carboxylate;hydrobromide

Specifications

300-08-3
White to Yellow
98% min. (Argentometry)
MFCD00039041
arecoline hydrobromide, arecoline bromide, taeniolin, arecoline hbr, methyl 1-methyl-1,2,5,6-tetrahydropyridine-3-carboxylate hydrobromide, arekolinhydrobromid german, unii-24s79b9cx7, methyl 1,2,5,6-tetrahydro-1-methylnicotinate hydrobromide, n-methyltetrahydronicotinic acid methyl ester hydrobromide
AXOJRQLKMVSHHZ-UHFFFAOYSA-N
methyl 1-methyl-3,6-dihydro-2H-pyridine-5-carboxylate;hydrobromide
9301
98+%
Arecoline hydrobromide
171°C to 174°C
Authentic
C8H14BrNO2
15, 768
Solubility in water: 1000g/L (20°C). Other solubilities: soluble in ethanol,slightly soluble in chloroform and ether
[H+].[Br-].COC(=O)C1=CCCN(C)C1
236.11
236.11
Powder
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

missing translation for 'einecsNumber' : 206-087-3

Product Identifier
  • Arecoline hydrobromide

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 3

Hazard Statement
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H301-Toxic if swallowed.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only