Learn More
Applied Biosystems™ MicroAmp™ 12-Cap Strip
Designed to fit on MicroAmp reaction tubes, tube strips, and 96-well plates
Brand: Applied Biosystems™ N8011534
Code : NEW
Additional Details : Weight : 0.50000kg
Description
MicroAmp™ Cap Strips have a domed profile and are suitable for end-point PCR. 12-cap strips available in clear or an assorted set of red, orange, blue, and green.
- Will not interfere with sample reading
- Provides tight seal to minimize evaporation
- Easy to apply and remove
- Use MicroAmp Caps Strips to seal tubes and reduce possibility of contamination in sample wells and help ensure reliable results
- Easy to Use
- Use MicroAmp Cap Installing Tool (sold separately) to push caps down and form tight seal
- When ready, just peel away caps using tabs at end
- Colored for Easy Organization
- Available in different colors to help track tubes during assay preparation
See user's manual or package insert for limited label license and trademark information.
PCR, PCR & Real-Time PCR, Real Time PCR (qPCR)
Specifications
Certified DNA/Rnase Free | |
1000 Strips (5 pk./200) | |
MicroAmp™ | |
Contains 1,000 MicroAmp™ 12-Cap Strips (5 packs of SKU# N8010534, Qty 200 per pack). Store at room temperature. | |
96 | |
12-Strip Cap |
Assorted | |
Thermal Cyclers | |
Polypropylene | |
Domed Caps | |
1,000 Strip Caps |