Learn More
AP5Z1/SPG48 Mouse, anti-Human, CoraLite 594, Clone: 1G1D7, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech CL594-66533-100UL
This item is not returnable.
View return policy
Description
AP5Z1, also known as SPG48 and KIAA0415, is a part of AP-5. Biallelic mutations in the AP5Z1 gene encoding the AP5Z1 subunit have been described in a small number of patients with hereditary spastic paraplegia (HSP). AP5Z1 physically interacts with other HSP proteins and that patient cells are sensitive to DNA damaging drugs.
Specifications
AP5Z1/SPG48 | |
Monoclonal | |
1000 μg/mL | |
Mouse | |
Primary | |
Human, Rat |
Immunofluorescence | |
1G1D7 | |
CoraLite 594 | |
100 μL | |
9907 | |
Liquid |
Product Content Correction
Product Title
AP5Z1/SPG48 Mouse, anti-Human, CoraLite 594, Clone: 1G1D7, Proteintech > 100 μL
Spot an opportunity for improvement?Share a Content Correction