missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Antithrombin III Mouse anti-Human, Mouse, Rat, Clone: 2B2E5, Proteintech
Mouse Monoclonal Antibody
£135.00 - £349.00
Specifications
Antigen | Antithrombin III |
---|---|
Clone | 2B2E5 |
Concentration | 0.62 mg/mL |
Applications | Western Blot, Immunocytochemistry, Immunohistochemistry (Paraffin), Immunofluorescence |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16897575
|
Proteintech
66052-1-IG-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16887575
|
Proteintech
66052-1-IG-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The protein encoded by this gene is a plasma protease inhibitor and a member of the serpin superfamily. This protein inhibits thrombin as well as other activated serine proteases of the coagulation system, and it regulates the blood coagulation cascade. The protein includes two functional domains: the heparin binding-domain at the N-terminus of the mature protein, and the reactive site domain at the C-terminus. The inhibitory activity is enhanced by the presence of heparin. More than 120 mutations have been identified for this gene, many of which are known to cause antithrombin-III deficiency.Specifications
Antithrombin III | |
0.62 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Antithrombin, Antithrombin III, AT3, ATIII, HEPARIN COFACTOR I, Serpin C1, SERPINC1 | |
SERPINC1 | |
IgG1 | |
Protein A | |
Antibody |
2B2E5 | |
Western Blot, Immunocytochemistry, Immunohistochemistry (Paraffin), Immunofluorescence | |
Unconjugated | |
Mouse | |
Rat, Mouse, Human | |
P01008, P32261 | |
11905, 304917, 462 | |
Antithrombin III Fusion Protein Ag9880 | |
Primary | |
-20°C | |
SERPINC1 |