Learn More
Antimony(III) n-butoxide, 99+%, Thermo Scientific™
Preparation of coatings, as a catalyst
£21.30 - £332.00
Chemical Identifiers
CAS | 2155-74-0 |
---|---|
Molecular Formula | C12H27O3Sb |
Molecular Weight (g/mol) | 341.105 |
MDL Number | MFCD00015238 |
InChI Key | YGBFTDQFAKDXBZ-UHFFFAOYSA-N |
Synonym | antimony iii butoxide, 1-butanol, antimony 3+ salt, antimony iii n-butoxide, tributoxystibine, antimony 3+ tributan-1-olate, acmc-20als1, antimony iii butan-1-olate, antimony 3+ ; butan-1-olate, antimony 3+ ion tris butan-1-olate, antimony 3+ tris butan-1-olate |
PubChem CID | 102217 |
IUPAC Name | antimony(3+);butan-1-olate |
SMILES | CCCC[O-].CCCC[O-].CCCC[O-].[Sb+3] |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
2155-74-0 | |
341.105 | |
YGBFTDQFAKDXBZ-UHFFFAOYSA-N | |
102217 | |
CCCC[O-].CCCC[O-].CCCC[O-].[Sb+3] |
C12H27O3Sb | |
MFCD00015238 | |
antimony iii butoxide, 1-butanol, antimony 3+ salt, antimony iii n-butoxide, tributoxystibine, antimony 3+ tributan-1-olate, acmc-20als1, antimony iii butan-1-olate, antimony 3+ ; butan-1-olate, antimony 3+ ion tris butan-1-olate, antimony 3+ tris butan-1-olate | |
antimony(3+);butan-1-olate |
Specifications
2155-74-0 | |
133°C to 135°C (0.4 mmHg) | |
C12H27O3Sb | |
UN3082 | |
antimony iii butoxide, 1-butanol, antimony 3+ salt, antimony iii n-butoxide, tributoxystibine, antimony 3+ tributan-1-olate, acmc-20als1, antimony iii butan-1-olate, antimony 3+ ; butan-1-olate, antimony 3+ ion tris butan-1-olate, antimony 3+ tris butan-1-olate | |
YGBFTDQFAKDXBZ-UHFFFAOYSA-N | |
antimony(3+);butan-1-olate | |
102217 | |
≥99% | |
Antimony (III) n-butoxide |
1.264 g/mL | |
>110°C (230°F) | |
MFCD00015238 | |
Moisture sensitive | |
Soluble in anhydrous butanol | |
CCCC[O-].CCCC[O-].CCCC[O-].[Sb+3] | |
341.105 | |
341.1 | |
Liquid |
Safety and Handling
GHS H Statement
H302-H332
Harmful if swallowed.
Harmful if inhaled.
P261-P264b-P270-P271-P301+P312-P304+P340-P312-P330-P501c
H302+H332
missing translation for 'dotInformation' : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCE, LIQUID, N.O.S.
missing translation for 'einecsNumber' : 218-456-6
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
Your input is important to us. Please complete this form to provide feedback related to the content on this product.