missing translation for 'onlineSavingsMsg'
Learn More
Learn More
WNT6 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA534326
Code : NEW
Additional Details : Weight : 0.01000kg
Description
Percent identity with other species by BLAST analysis: Human, Gorilla, Gibbon, Monkey, Mouse, Rat, Hamster, Elephant, Panda, Bovine, Dog, Horse, Rabbit (100%).
WNT6 Polyclonal antibody specifically detects WNT6 in Bovine, Equine, Hamster, Human, Mouse, Rabbit, Rat samples. It is validated for Immunohistochemistry (Paraffin).Specifications
WNT6 | |
Polyclonal | |
Unconjugated | |
WNT6 | |
AA409270; Protein Wnt-6; wingless-related MMTV integration site 6; wingless-related MMTV integration site 6 homolog; wingless-type MMTV integration site family member 6; wingless-type MMTV integration site family, member 6; Wnt family member 6; WNT6; Wnt-6 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
100058824, 100348986, 100751289, 22420, 316526, 505075, 7475 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.69 mg/mL | |
PBS with 0.1% sodium azide | |
P22727, Q9Y6F9 | |
WNT6 | |
Synthetic 20 amino acid peptide from N-terminus of human WNT6. | |
50 μg | |
Primary | |
Bovine, Equine, Hamster, Human, Mouse, Rabbit, Rat | |
Antibody | |
IgG |