Learn More
UXT Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA543112
Code : NEW
Additional Details : Weight : 0.00010kg
Description
Peptide sequence: MATPPKRRAV EATGEKVLRY ETFISDVLQR DLRKVLDHRD KVYEQLAKYL Sequence homology: Cow: 93%; Dog: 100%; Guinea Pig: 93%; Horse: 100%; Human: 100%; Rabbit: 86%; Rat: 93%
UXT Polyclonal antibody specifically detects UXT in Human samples. It is validated for Western Blot.Specifications
UXT | |
Polyclonal | |
Unconjugated | |
UXT | |
0910002B17Rik; androgen receptor trapped clone 27 protein; ART-27; HSPC024; Protein UXT; RP1-212G6.2; SKP2-associated alpha PFD 1; STAP1; ubiquitously expressed prefoldin like chaperone; ubiquitously expressed transcript; ubiquitously expressed transcript protein; ubiquitously-expressed transcript; ubiquitously-expressed, prefoldin-like chaperone; Unknown (protein for MGC:133475); UXT | |
Rabbit | |
Protein A | |
RUO | |
8409 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q9UBK9 | |
UXT | |
Synthetic peptide directed towards the N-terminal of human UXT | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |