Learn More
USP7 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA534911
Code : NEW
Additional Details : Weight : 0.01000kg
Description
Recommended positive controls: 293T, A431, HeLa, HepG2, USP7 transfected 293T lysate, NIH-3T3, JC, PC-12. Predicted reactivity: Mouse (97%), Rat (97%), Xenopus laevis (93%), Pig (97%), Chicken (93%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
USP7 Polyclonal antibody specifically detects USP7 in Human, Mouse, Rat samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot.Specifications
USP7 | |
Polyclonal | |
Unconjugated | |
USP7 | |
2210010O09Rik; AA409944; AA617399; AU019296; AW548146; C80752; deubiquitinating enzyme 7; Hausp; Herpes virus-associated ubiquitin-specific protease; herpes-virus-associated ubiquitin-specific protease; herpesvirus-associated ubiquitin-specific protease; mHAUSP; protease; rHAUSP; TEF1; Ubiquitin carboxyl-terminal hydrolase 7; ubiquitin specific peptidase 7; ubiquitin specific peptidase 7 (herpes virus-associated); ubiquitin specific protease 7; ubiquitin specific protease 7 (herpes virus-associated); ubiquitin thioesterase 7; ubiquitin thiolesterase 7; ubiquitin-specific-processing protease 7; UBP; USP7 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
252870, 360471, 7874 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.94 mg/mL | |
PBS with 20% glycerol and 0.025% ProClin 300; pH 7 | |
Q4VSI4, Q6A4J8, Q93009 | |
USP7 | |
Recombinant fragment corresponding to a region within amino acids 1 and 221 of USP7 (Uniprot ID#Q93009) | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |