missing translation for 'onlineSavingsMsg'
Learn More
Learn More
USP38 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA539512
Code : NEW
Additional Details : Weight : 0.00900kg
Specifications
USP38 | |
Polyclonal | |
Unconjugated | |
USP38 | |
KIAA1891 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
84640 | |
-20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q8NB14 | |
USP38 | |
A synthetic peptide derived from the internal region of human USP38 | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |