missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ULK1 Polyclonal Antibody, Invitrogen™
Sheep Polyclonal Antibody
Brand: Invitrogen
Code : LD
Additional Details : Weight : 0.01000kg
Description
Glycerol (1:1) may be added for additional stability. Reconstitute with 150 μL of distilled water. Centrifuge to remove insoluble material.
Involved in axon growth. Plays an essential role in neurite extension of cerebellar granule cells. Interacts with GABARAP and GABARAPL2.Specifications
ULK1 | |
Polyclonal | |
ULK1 | |
Unc-51-like kinase 1, serine/threonine-protein kinase Unc51.1 KIAA0722, ULK1, ATG1, UNC51, Unc51.1, unc-51-like kinase 1 (C. elegans), FLJ38455 | |
Sheep | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Immunohistochemistry, Western Blot | |
Unconjugated | |
O75385, O70405 | |
ULK1 | |
A synthetic peptide from internal region of human ULK1 | |
RUO | |
22241, 360827, 8408 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Lyophilized |