missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TDP-43 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA517011
Code : LE
Additional Details : Weight : 0.01000kg
Description
It is not recommended to aliquot this antibody.
TDP43 was first identified as a novel cellular protein that binds to HIV-1 virus TAR DNA sequence motifs and acts a transcriptional repressor to the HIV-1 LTR. Later experiments revealed that TDP43 also regulates the splicing of exon 9 of the cystic fibrosis transmembrane conductance regular (CFTR), most likely through the association with the UG repeats at the 3"e;e; splice site. A hyperphosphorylated, ubiquitinated, and cleaved form of TDP43 known as pathologic TDP43 is the major disease protein in ubiquitin-positive, tau-, and alpha-synuclein-negative frontotemporal dementia (FLTD-U). TDP43 is not related to TRBP1, and RNA binding protein that binds HIV-1 TAR RNA sequences.Specifications
TDP-43 | |
Polyclonal | |
Unconjugated | |
TARDBP | |
1190002A23Rik; ALS10; C85084; hypothetical protein; OTTHUMP00000002171; OTTHUMP00000002173; RCJMB04_5g9; RP4-635E18.2; TAR DNA binding protein; TAR DNA binding protein L homeolog; TAR DNA-binding protein 43; TAR DNA-binding protein-43; tardbp; tardbp.L; Tdp43; TDP-43; wu:fb77f02; wu:fc52g10; XELAEV_18035188mg | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
230908, 23435, 298648 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunoprecipitation, Western Blot | |
14 μg/mL | |
0.01M HEPES with 0.15M NaCl, 100μg/mL BSA, 50% glycerol and no preservative; pH 7.5 | |
Q13148, Q921F2 | |
TARDBP | |
Synthetic peptide corresponding to residues surrounding Gly400 of human TDP43 | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |