Learn More
TCP-1 beta Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA522221
Code : LD
Additional Details : Weight : 0.01000kg
Description
Recommended positive controls: 293T, A431, H1299, HeLa, HepG2, Molt-4, Raji, mouse brain. Predicted reactivity: Mouse (97%), Rat (97%), Zebrafish (87%), Xenopus laevis (93%), Pig (100%), Rabbit (100%), Chicken (93%), Rhesus Monkey (100%), Chimpanzee (100%), Bovine (98%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
TCP-1 beta is a molecular chaperone that is member of the chaperonin containing TCP1 complex (CCT), also known as the TCP1 ring complex (TRiC). The CCT complex consists of two identical stacked rings, each containing eight different proteins. Unfolded polypeptides enter the central cavity of the complex and are folded in an ATP-dependent manner. The complex folds various proteins, including actin and tubulin. Alternate transcriptional splice variants of the TCP-1 beta gene have been observed but not thoroughly characterized. A disease associated with TCP-1 beta protein dysfunction include epididymo-orchitis.Specifications
TCP-1 beta | |
Polyclonal | |
Unconjugated | |
CCT2 | |
99D8.1; AI528772; c-cpn; CCT; Cct1; CCT2; Ccta; CCTB; CCT-beta; chaperonin containing t-complex polypeptide 1, beta subunit; chaperonin containing t-complex polypeptide 1, subunit 2; chaperonin containing TCP1 subunit 2; chaperonin containing Tcp1, subunit 2 (beta); chaperonin subunit 2 (beta); chaperonin-containing T-complex polypeptide beta subunit; epididymis secretory sperm binding protein Li 100n; HEL-S-100n; hypothetical protein LOC505313; p63; PRO1633; T-complex protein 1 subunit beta; T-complex protein 1, beta subunit; Tcp-1; TCP-1-beta; Tp63; TRic | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
10576, 12461 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
P78371, P80314 | |
CCT2 | |
Recombinant fragment corresponding to a region within amino acids 277 and 503 of TCP1 beta (Uniprot ID#P78371) | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |