Learn More
TBX2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA540713
Code : NEW
Additional Details : Weight : 0.00010kg
Description
Peptide sequence: AEIAQAEEQA RKRQEEREKE AAEQAERSQS SIVPEEEQAA NKGEEKKDDE Sequence homology: Cow: 92%; Dog: 85%; Guinea Pig: 85%; Horse: 92%; Human: 100%; Mouse: 77%; Pig: 92%; Rabbit: 92%; Rat: 85%
TBX2 Polyclonal antibody specifically detects TBX2 in Human samples. It is validated for Western Blot.Specifications
TBX2 | |
Polyclonal | |
Unconjugated | |
TBX2 | |
FLJ10169; LOW QUALITY PROTEIN: T-box transcription factor TBX2; T-box 2; T-box 2 protein; T-box 2 transcription factor; T-box protein 2; T-box transcription factor 2; T-box transcription factor TBX2; TBX2 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
6909 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q13207 | |
TBX2 | |
Synthetic peptide directed towards the middle region of human TBX2 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |