Learn More
TAAR9 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA533070
Code : PN
Additional Details : Weight : 0.01000kg
Description
In IHC applications, antigen retrieval using a citrate buffer (pH 6.0) is recommended. Percent identity with other species by BLAST analysis: Human (100%) Hamster (92%) Galago, Mouse, Rat, Bovine, Pig (85%).
TAAR9 is a member of a large family of rhodopsin G protein-coupled receptors (GPCRs, or GPRs). GPCRs contain 7 transmembrane domains and transduce extracellular signals through heterotrimeric G proteins.Specifications
TAAR9 | |
Polyclonal | |
Unconjugated | |
Q96RI9 | |
TAAR9 | |
Synthetic 13 amino acid peptide from 3rd extracellular domain of human TAAR9. | |
50 μg | |
Primary | |
Human | |
Antibody | |
IgG |
ELISA, Immunohistochemistry (Paraffin) | |
0.8 mg/mL | |
TAAR9 | |
TA3, TAR3, TAR9, TRAR3, trace amine receptor 3, trace amine receptor 9, trace amine-associated receptor 9 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
134860 | |
Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C | |
Liquid |