missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SRSF4 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA538663
Code : NEW
Additional Details : Weight : 0.00900kg
Description
Serine/arginine-rich protein 75 (SRp75) is encoded by the SRSF4 gene. SRp75 belongs to the arginine/serine-rich factor family and likely plays a role in alternative mRNA splicing.The family of SR factors all contain one or more RNA recognition motifs (RRM) and an arginine/ serine (RS)-rich domain. They are not only essential for constitutive splicing but also regulate splicing in a concentration-dependent manner by influencing the selection of alternative splice sites. The majority of SR proteins, including SC35 and SRp40, are confined to the nucleus, while SF2/ASF, SRp20, and 9G8 are continuously shuttled between the nucleus and the cytoplasm and contribute to mRNA transport. The activity of SR proteins in regulated splicing is antagonized by members of the hnRNP A/B family of proteins, which induce drastic shifts in the selection of splicing sites.Specifications
SRSF4 | |
Polyclonal | |
Unconjugated | |
SRSF4 | |
arginine/serine-rich 4, Pre mRNA splicing factor SRP75, Pre-mRNA-splicing factor SRP75, Serine/arginine-rich splicing factor 4, Splicing factor, Splicing factor, arginine/serine rich 4, SRP001LB, SRP75, Srsf4 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
57317, 6429 | |
-20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q08170, Q8VE97 | |
SRSF4 | |
A synthetic peptide derived from the internal region of human SFRS4 | |
100 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |