missing translation for 'onlineSavingsMsg'
Learn More

SRD5A1, Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™

Rabbit polyclonal antibody raised against a full-length human SRD5A1 protein.

Brand:  Abnova H00006715-D01P.100ug

Additional Details : Weight : 0.00010kg

Product Code. 16165825

  • £286.00 / 100µg
Estimated Shipment: 10-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Steroid 5-alpha-reductase (EC 1.3.99.5) catalyzes the conversion of testosterone into the more potent androgen, dihydrotestosterone (DHT). Also see SRD5A2 (MIM 607306).[supplied by OMIM

Sequence: MATATGVAEERLLAALAYLQCAVGCAVFARNRQTNSVYGRHALPSHRLRVPARAAWVVQELPSLALPLYQYASESAPRLRSAPNCILLAMFLVHYGHRCLIYPFLMRGGKPMPLLACTMAIMFCTCNGYLQSRYLSHCAVYADDWVTDPRFLIGFGLWLTGMLINIHSDHILRNLRKPGDTGYKIPRGGLFEYVTAANYFGEIMEWCGYALASWSVQGAAFAFFTFCFLSGRAKEHHEWYLRKFEEYPKFRKIIIPFLF
Specifications

Specifications

SRD5A1
Polyclonal
Rabbit polyclonal antibody raised against a full-length human SRD5A1 protein.
SRD5A1
SRD5A1
SRD5A1 (NP_001038.1, 1 a.a. ∼ 259 a.a) full-length human protein.
100 μg
Primary
Human
Antibody
IgG
Western Blot
Unconjugated
PBS with no preservative; pH 7.4
NM_001047.2
Rabbit
Affinity chromatography
RUO
6715
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers