missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SPINK1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA540796
Code : NEW
Additional Details : Weight : 0.00010kg
Description
Peptide sequence: KVTGIFLLSA LALLSLSGNT GADSLGREAK CYNELNGCTK IYDPVCGTDG Sequence homology: Human: 100%; Rabbit: 93%
SPINK1 Polyclonal antibody specifically detects SPINK1 in Human samples. It is validated for Western Blot.Specifications
SPINK1 | |
Polyclonal | |
Unconjugated | |
SPINK1 | |
calcium transport inhibitor; Caltrin; ISK1; P12; Pancreatic secretory trypsin inhibitor; pancreatic secretory trypsin inhibitor II; pancreatic secretory trypsin inhibitor type II (PSTI-II); PCTT; prostatic secretory glycoprotein; PSTI; PSTI-II; serine peptidase inhibitor Kazal type 1; serine peptidase inhibitor, Kazal type 1; serine peptidase inhibitor, Kazal type 3; serine protease inhibitor Kazal-type 1; Serine protease inhibitor Kazal-type 1-like; Serine protease inhibitor Kazal-type 3; serine protease inhibitor, Kazal type 1; serine protease inhibitor, Kazal type 3; Spink 3; Spink1; Spink1l; Spink3; TATI; TCP; Tumor-associated trypsin inhibitor | |
Rabbit | |
Affinity Chromatography | |
RUO | |
6690 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
P00995 | |
SPINK1 | |
Synthetic peptide directed towards the N-terminal of human SPINK1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |