Learn More
SPHKAP Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA527581
Code : PN
Additional Details : Weight : 0.01000kg
Description
Recommended positive controls: NT2D1, IMR32, MCF-7, SK-N-SH, IMR32, SK-N-AS. Predicted reactivity: Mouse (93%), Rat (93%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Anchoring protein that binds preferentially to the type I regulatory subunit of c-AMP-dependent protein kinase (PKA type I) and targets it to distinct subcellular compartments. May act as a converging factor linking cAMP and sphingosine signaling pathways. Plays a regulatory role in the modulation of SPHK1. [UniProt]Specifications
SPHKAP | |
Polyclonal | |
Unconjugated | |
SPHKAP | |
A-kinase anchor protein SPHKAP; KIAA1678; SKIP; Sphingosine kinase type 1-interacting protein; SPHK1 (sphingosine kinase type 1) interacting protein; SPHK1 interactor, AKAP domain containing; SPHK1-interactor and AKAP domain-containing protein; SPHKAP | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
80309 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
Q2M3C7 | |
SPHKAP | |
Recombinant protein fragment corresponding to a region within amino acids 1 and 219 of Human SPHKAP | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |