missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SPAM1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA545549
Code : NEW
Additional Details : Weight : 0.00010kg
Description
Peptide sequence: CYSTLSCKEK ADVKDTDAVD VCIADGVCID AFLKPPMETE EPQIFYNASP Sequence homology: Human: 100%
SPAM1 Polyclonal antibody specifically detects SPAM1 in Human samples. It is validated for Western Blot.Specifications
SPAM1 | |
Polyclonal | |
Unconjugated | |
SPAM1 | |
AV039194; epididymis secretory sperm binding protein Li 96n; HEL-S-96n; HYA1; HYAL1; HYAL3; HYAL5; hyal-PH20; Hyaluronidase PH-20; hyaluronoglucosaminidase PH-20; MGC26532; PH20; Ph-20; PH-20 hyaluronidase; SPAG15; Spam; SPAM1; Sperm adhesion molecule 1; sperm adhesion molecule 1 (PH-20 hyaluronidase, zona pellucida binding); sperm surface antigen 2B1; Sperm surface protein PH-20 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
6677 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
P38567 | |
SPAM1 | |
Synthetic peptide directed towards the C-terminal of human SPAM1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |