missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sclerostin Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Brand: Invitrogen PA547134
Code : NEW
Additional Details : Weight : 0.00010kg
Description
In direct ELISAs, approximately 15% cross-reactivity with recombinant mouse SOST is observed. Reconstitute at 0.2 mg/mL in sterile PBS.
Sclerostin Polyclonal antibody specifically detects Sclerostin in Human samples. It is validated for Western Blot.Specifications
Sclerostin | |
Polyclonal | |
Unconjugated | |
SOST | |
5430411E23Rik; CDD; DAND6; sclerosteosis; sclerostin; SOST; SOST1; UNQ2976/PRO7455/PRO7476; VBCH | |
Goat | |
Antigen affinity chromatography | |
RUO | |
50964 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot | |
0.2 mg/mL | |
PBS with 5% trehalose and No Preservative | |
Q9BQB4 | |
SOST | |
Mouse myeloma cell line NS0-derived recombinant human SOST Gln24-Tyr213 | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |