missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SNX27 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA523025
Code : LD
Additional Details : Weight : 0.01000kg
Description
This antibody is predicted to react with rat, bovine and zebrafish based on 100% sequence homology.
SNX27 encodes a member of the sorting nexin family, a diverse group of cytoplasmic and membrane-associated proteins involved in endocytosis of plasma membrane receptors and protein trafficking through these compartments. All members of this protein family contain a phosphoinositide binding domain (PX domain). A highly similar protein in mouse is responsible for the specific recruitment of an isoform of serotonin 5-hydroxytryptamine 4 receptor into early endosomes, suggesting the analogous role for the human protein.Specifications
SNX27 | |
Polyclonal | |
Unconjugated | |
SNX27 | |
5730552M22Rik; ESTM45; ESTM47; KIAA0488; MAP-responsive gene protein; methamphetamine (MAP) responsive transcript 1; methamphetamine-responsive transcript 1; methamphetamine-responsive transcript 1 protein; Mrt1; MY014; PDZ protein Mrt1; PDZ-protein Mrt1; R75405; RP11-98D18.12-005; SNX27; sorting nexin family member 27; sorting nexin-27 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
260323, 76742, 81609 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
1 mg/mL | |
PBS with 30% glycerol and 0.1% sodium azide | |
Q3UHD6, Q8K4V4, Q96L92 | |
SNX27 | |
Synthetic peptide made to an internal portion of the human protein (within residues 450-500). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |