Learn More
SNIP1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA529412
Code : PN
Additional Details : Weight : 0.01000kg
Description
Recommended positive controls: 293T, A431, Jurkat, Raji. Predicted reactivity: Mouse (86%), Rat (87%), Rhesus Monkey (97%), Bovine (88%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Smad-binding peptide aptamers can be developed to selectively inhibit TGF-beta-induced gene expression.Specifications
SNIP1 | |
Polyclonal | |
Unconjugated | |
SNIP1 | |
2410133M08Rik; dJ423B22.2; FHA domain-containing protein SNIP1; PMRED; RP3-423B22.3; Smad nuclear interacting protein; smad nuclear interacting protein 1; Smad nuclear interacting protein 1 Snip1; smad nuclear-interacting protein 1; Snip1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
313588, 76793, 79753 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
Q5M9G6, Q8BIZ6, Q8TAD8 | |
SNIP1 | |
Recombinant fragment corresponding to a region within amino acids 41 and 363 of Human SNIP1 | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |