Learn More
SLC27A4 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA542446
Code : NEW
Additional Details : Weight : 0.00010kg
Description
Peptide sequence: YKFQKTELRK EGFDPAIVKD PLFYLDAQKG RYVPLDQEAY SRIQAGEEKL Sequence homology: Cow: 86%; Dog: 86%; Guinea Pig: 93%; Horse: 93%; Human: 100%; Mouse: 93%; Rat: 93%
SLC27A4 Polyclonal antibody specifically detects SLC27A4 in Human samples. It is validated for Western Blot.Specifications
SLC27A4 | |
Polyclonal | |
Unconjugated | |
SLC27A4 | |
ACSVL4; BB144259; Fatp4; FATP-4; fatty acid transport protein 4; IPS; Long-chain fatty acid transport protein 4; SLC27A4; solute carrier family 27 (fatty acid transporter), member 4; solute carrier family 27 member 4 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
10999 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q6P1M0 | |
SLC27A4 | |
Synthetic peptide directed towards the middle region of human SLC27A4 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |