Learn More
SLC25A12 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA542430
Code : NEW
Additional Details : Weight : 0.00010kg
Description
Peptide sequence: GLKPAGSEPT PKSRIADLPP ANPDHIGGYR LATATFAGIE NKFGLYLPKF Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 93%; Horse: 93%; Human: 100%; Mouse: 100%; Pig: 100%; Rabbit: 100%; Rat: 100%
SLC25A12 Polyclonal antibody specifically detects SLC25A12 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Specifications
SLC25A12 | |
Polyclonal | |
Unconjugated | |
SLC25A12 | |
2610002D09Rik; AGC1; AI839531; araceli hiperlarga; ARALAR; Aralar1; B230107K20Rik; BB129864; calcium binding mitochondrial carrier superfamily member Aralar1; calcium-binding mitochondrial carrier protein Aralar1; Calcium-binding mitochondrial carrier protein Aralar1-like protein; mitochondrial aspartate glutamate carrier 1; RGD1561141; SLC25A12; solute carrier family 25 (aspartate/glutamate carrier), member 12; solute carrier family 25 (mitochondrial carrier, Aralar), member 12; solute carrier family 25 member 12; solute carrier family 25, member 12 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
8604 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
O75746 | |
SLC25A12 | |
Synthetic peptide directed towards the middle region of human SLC25A12 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |