missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLAIN1 Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Brand: Invitrogen PA547760
Code : NEW
Additional Details : Weight : 0.00010kg
Description
Reconstitute at 0.2 mg/mL in sterile PBS.
SLAIN1 Polyclonal antibody specifically detects SLAIN1 in Human, Mouse samples. It is validated for Immunocytochemistry, Western Blot.Specifications
SLAIN1 | |
Polyclonal | |
Unconjugated | |
SLAIN1 | |
9630044O09Rik; AA675320; AW742596; C13orf32; RGD1308626; SLAIN motif family member 1; SLAIN motif family, member 1; SLAIN motif-containing protein 1; SLAIN1 | |
Goat | |
Antigen affinity chromatography | |
RUO | |
105439, 122060 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunocytochemistry, Western Blot | |
0.2 mg/mL | |
PBS with 5% trehalose and No Preservative | |
Q68FF7, Q8ND83 | |
SLAIN1 | |
E. coli-derived recombinant mouse SLAIN1 Met262-Gln483 | |
100 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |