missing translation for 'onlineSavingsMsg'
Learn More

Donkey anti-Sheep IgG (H+L) Secondary Antibody, Invitrogen™

Donkey Polyclonal Secondary Antibody

Brand:  Invitrogen A16043

Product Code. 15313886

  • £89.75 / 2mg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

The sensitivity of each lot of antibody is confirmed using ELISA. The specificity of each lot of antibody is confirmed by immunoelectrophoresis (IEP).

Anti-Sheep secondary antibodies are affinity-purified antibodies with well-characterized specificity for ovine (sheep) immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Sheep IgG (H+L)
Polyclonal
Unconjugated
Donkey
Affinity chromatography
RUO
Ovine
Liquid
ELISA
> 4.5 mg/mL
PBS with 0.05% sodium azide; pH 7.2
Gamma Immunoglobins Heavy and Light chains
2 mg
Secondary
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers