missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SHANK1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA146252
Code : LD
Additional Details : Weight : 0.01000kg
Description
Shank is a novel family of postsynaptic density proteins (PSD) that bind to PSD-95-associated protein GKAP. Its putative function is to act as a scaffold protein in the PSD to cross-link NMDA receptors/PSD-95 complexes and couple them to regulators of thSpecifications
SHANK1 | |
Polyclonal | |
Unconjugated | |
Q9Y566, D3YZU1, Q9WV48 | |
SHANK1 | |
Synthetic peptide: ERLLREYPQSFEKGV, containing C terminal amino acids 135-149 of Human SHANK1 | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Western Blot | |
1 mg/mL | |
SHANK1 | |
SPANK-1, SSTRIP, synamon | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
243961, 50944, 78957 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |