Learn More
Goat anti-Rat IgM (Heavy chain) Secondary Antibody, Alexa Fluor™ 647, Invitrogen™
Goat Polyclonal Secondary Antibody
Brand: Invitrogen A21248
Code : 50
Additional Details : Weight : 0.00450kg
Description
This goat anti-Rat antibody has been purified by rat IgM-affinity chromatography and reacts specifically with IgM heavy chains (μ chains) and not with immunoglobulin light chains from rat, as determined by ELISA. Product will be shipped at Room Temperature.
Rat IgM (Heavy chain) Polyclonal antibody specifically detects Rat IgM (Heavy chain) in Rat samples. It is validated for Immunocytochemistry, Immunohistochemistry.Specifications
Rat IgM (Heavy chain) | |
Polyclonal | |
Alexa Fluor 647 | |
Goat | |
purified | |
RUO | |
Rat | |
Liquid |
Immunocytochemistry, Immunohistochemistry | |
2 mg/mL | |
PBS with 5mM sodium azide; pH 7.5 | |
Rat Mu immunonglobin | |
500 μg | |
Secondary | |
4° C, store in dark | |
IgG |