missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Goat anti-Rat IgM Cross-Adsorbed Secondary Antibody, DyLight™ 680, Invitrogen™
Goat Polyclonal Secondary Antibody
Brand: Invitrogen SA510014
Code : PN
Additional Details : Weight : 0.01000kg
Description
This antibody is cross-adsorbed and exhibits minimum reactivity to human and mouse.
Anti-Rat secondary antibodies are affinity-purified antibodies with well-characterized specificity for rat immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.Specifications
Rat IgM Cross-Adsorbed | |
Polyclonal | |
DyLight 680 | |
Goat | |
Antigen affinity chromatography | |
RUO | |
Rat | |
Liquid |
Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot | |
0.5 mg/mL | |
PBS with 0.2% BSA and 0.09% sodium azide; pH 6.8 to 7.4 | |
Rat IgM | |
500 μg | |
Secondary | |
4° C | |
IgG |