missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Goat anti-Rat IgG (H+L) Cross-Adsorbed Secondary Antibody, PE, Invitrogen™
Goat Polyclonal Secondary Antibody
Brand: Invitrogen A10545
Code : 50
Additional Details : Weight : 0.00450kg
Description
Anti-Rat secondary antibodies are affinity-purified antibodies with well-characterized specificity for rat immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.Specifications
Rat IgG (H+L) Cross-Adsorbed | |
Polyclonal | |
PE | |
Goat | |
purified | |
RUO | |
Rat | |
Liquid |
Immunocytochemistry | |
1 mg/mL | |
PBS with 0.1% sodium azide; pH 7.5 | |
Gamma Immunoglobins Heavy and Light chains | |
250 μL | |
Secondary | |
4° C | |
IgG |