missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-RARA (Ser77) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA539870
Code : NEW
Additional Details : Weight : 0.00900kg
Description
This gene represents a nuclear retinoic acid receptor. The encoded protein, retinoic acid receptor alpha, regulates transcription in a ligand-dependent manner. This gene has been implicated in regulation of development, differentiation, apoptosis, granulopoeisis, and transcription of clock genes. Translocations between this locus and several other loci have been associated with acute promyelocytic leukemia. Alternatively spliced transcript variants have been found for this locus.Specifications
RARA | |
Polyclonal | |
Unconjugated | |
RARA | |
RARA, RAR, NR1B1, retinoic acid receptor, alpha, Retinoic acid receptor, alpha polypeptide, nucleophosmin-retinoic acid receptor alpha fusion protein NPM-RAR long form, nucleophosmin-retinoic acid receptor alpha fusion protein NPM-RAR short form | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
19401, 24705, 5914 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
P10276, P11416 | |
RARA | |
A synthetic phosphopeptide derived from human Retinoic Acid Receptor alpha around the phosphorylation site of Ser77 (P-P-SP-P-P) | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |