missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PLRG1 Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Brand: Invitrogen PA517990
Code : A0
Additional Details : Weight : 0.01000kg
Description
This antibody is predicted to react with bovine, canine, human, porcine and rat based on sequence homology.
PLRG1 Polyclonal antibody specifically detects PLRG1 in Human, Mouse samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Specifications
PLRG1 | |
Polyclonal | |
Unconjugated | |
PLRG1 | |
AA958940; C80566; Cwc1; MGC110980; MGC137269 protein; pleiotropic regulator 1; pleiotropic regulator 1 (PRL1 homolog, Arabidopsis); pleiotropic regulator 1 homolog; pleiotropic regulator 1, PRL1 homolog (Arabidopsis); pleiotropic regulator 1-like protein; PLRG1; PRL1; PRP46; PRPF46; Tango4; transport and golgi organization 4 homolog | |
Goat | |
Antigen affinity chromatography | |
RUO | |
53317, 5356 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
O43660, Q922V4 | |
PLRG1 | |
Synthetic peptide sequence (PVSWKPEIIKRKRF) corresponding to the C-terminus amino acids of PLRG1 | |
100 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |