missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Plexin-A1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA517147
Code : LE
Additional Details : Weight : 0.01000kg
Description
It is not recommended to aliquot this antibody.
Plexin-A1 Polyclonal antibody specifically detects Plexin-A1 in Human, Rat samples. It is validated for Immunoprecipitation, Western Blot.Specifications
Plexin-A1 | |
Polyclonal | |
0.01M HEPES with 0.15M NaCl, 100μg/mL BSA, 50% glycerol and no preservative; pH 7.5 | |
Q9UIW2 | |
PLXNA1 | |
Synthetic peptide surrounding Gln1875 of human Plexin A1 | |
100 μL | |
Primary | |
Human, Rat | |
Antibody | |
IgG |
Immunoprecipitation, Western Blot | |
Unconjugated | |
PLXNA1 | |
2600013D04Rik; Kiaa4053; mKIAA4053; NOV; NOVP; Plex 1; PlexA1; plexin 1; plexin A1; plexin-1; plexin-A1; PLXN1; Plxna1; RGD1560871; semaphorin receptor NOV | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
362398, 5361 | |
-20°C | |
Liquid |