missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pleckstrin Monoclonal Antibody (2D8), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA519629
Code : NEW
Additional Details : Weight : 0.00010kg
Description
Peptide Sequence: PETIDLGALY LSMKDTEKGI KELNLEKDKK IFNHCFTGNC VIDWLVSNQS VRNRQEGLMI ASSLLNEGYL QPAGDMSKSA VDGTAENPFL DNPDAFYYFP DSGFFCEENS
Pleckstrin Monoclonal antibody specifically detects Pleckstrin in Human samples. It is validated for ELISA, Immunocytochemistry, Western Blot.Specifications
Pleckstrin | |
Monoclonal | |
0.5 mg/mL | |
PBS with no preservative; pH 7.4 | |
P08567 | |
PLEK | |
PLEK (AAH18549, 121 a.a. ∽ 230 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2a κ |
ELISA, Immunocytochemistry, Western Blot | |
2D8 | |
Unconjugated | |
PLEK | |
2010300B13Rik; FLJ27168; P47; platelet 47 kDa protein; Pleckstrin; Plek | |
Mouse | |
Affinity chromatography | |
RUO | |
5341 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |