missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospholamban Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA526004
Code : LD
Additional Details : Weight : 0.01000kg
Description
Phospholamban Polyclonal antibody specifically detects Phospholamban in Human, Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot.Specifications
Phospholamban | |
Polyclonal | |
Unconjugated | |
PLN | |
cardiac CMH18; Cardiac phospholamban; CMD1P; CMH18; phospholamban; Plb; Plm; PLN; unnamed protein product | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
18821, 5350 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
0.5 mg/mL | |
PBS with 0.09% sodium azide | |
P26678, P61014 | |
PLN | |
KLH conjugated synthetic peptide between 1-30 amino acids from human Phospholamban | |
400 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |