missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Platelet Monoclonal Antibody (AIP21), Invitrogen™
Rat Monoclonal Antibody
Brand: Invitrogen MA170084
Code : LD
Additional Details : Weight : 0.01000kg
Description
MA1-70084 detects Platelet in mouse samples. MA1-70084 has been successfully used in flow cytometry procedures. The MA1-70084 immunogen is platelets.
Platelet Monoclonal antibody specifically detects Platelet in Mouse samples. It is validated for Flow Cytometry.Specifications
Platelet | |
Monoclonal | |
1 mg/mL | |
PBS with 0.02% sodium azide | |
Rat | |
Protein G | |
RUO | |
Mouse | |
Antibody | |
IgM |
Flow Cytometry | |
AIP21 | |
Unconjugated | |
Thrombocytes | |
Platelet. | |
250 μg | |
Primary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |