missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PKD2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen OSP00018W
Code : LD
Additional Details : Weight : 0.01000kg
Description
This antibody is directed against the unphosphorylated form of PKD2, and the cross-reactivity with phosphorylated forms has not yet been determined.
This gene encodes a member of the polycystin protein family. The encoded protein contains multiple transmembrane domains, and cytoplasmic N- and C-termini. The protein may be an integral membrane protein involved in cell-cell/matrix interactions. The encoded protein may function in renal tubular development, morphology, and function, and may modulate intracellular calcium homoeostasis and other signal transduction pathways. This protein interacts with polycystin 1 to produce cation-permeable currents. Mutations in this gene have been associated with autosomal domit polycystic kidney disease.Specifications
PKD2 | |
Polyclonal | |
PKD2 | |
PKD2, Polycystin-2, Autosomal domit polycystic kidney disease type II protein, Polycystwin, R48321 | |
Rabbit | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Immunohistochemistry, Western Blot | |
Unconjugated | |
Q13563 | |
PKD2 | |
A synthetic peptide from the 4th cytoplasmic loop of human Polycystic kidney disease 2 protein | |
RUO | |
5311 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Lyophilized |