missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pirin Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Brand: Invitrogen PA518291
Code : A0
Additional Details : Weight : 0.01000kg
Description
This antibody is predicted to react with mouse based on sequence homology.
Pirin Polyclonal antibody specifically detects Pirin in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Specifications
Pirin | |
Polyclonal | |
Unconjugated | |
PIR | |
2310042L19Rik; iron-binding nuclear protein; PIR; Pirin; pirin (iron-binding nuclear protein); pirin like; pirinin; Pirnl; Probable quercetin 2,3-dioxygenase PIR; probable quercetinase | |
Goat | |
Antigen affinity chromatography | |
RUO | |
8544 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
O00625 | |
PIR | |
Synthetic peptide sequence (GSSKKVTLSVLSREC) corresponding to the N-terminus amino acids of PIR | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |