missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PILRA Polyclonal Antibody, Invitrogen™
Sheep Polyclonal Antibody
Brand: Invitrogen PA547843
Code : NEW
Additional Details : Weight : 0.00010kg
Description
Reconstitute in sterile PBS to a final concentration of 0.2 mg/mL.
PILRA Polyclonal antibody specifically detects PILRA in Human samples. It is validated for Flow Cytometry.Specifications
PILRA | |
Polyclonal | |
Unconjugated | |
PILRA | |
AV021745; cell surface receptor FDF03; FDF03; Inhibitory receptor PILR-alpha; paired immunoglobin like type 2 receptor alpha; paired immunoglobin-like type 2 receptor alpha; paired immunoglobulin-like type 2 receptor alpha; Pilra | |
Sheep | |
Antigen affinity chromatography | |
RUO | |
29992 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Flow Cytometry | |
0.2 mg/mL | |
PBS with 5% trehalose and No Preservative | |
Q9UKJ1 | |
PILRA | |
Mouse myeloma cell line NS0-derived recombinant human PILR-alpha Gln20-Thr196 | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |