missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-PIKFYVE (Ser307) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA538808
Code : NEW
Additional Details : Weight : 0.00900kg
Description
Phospho-PIKFYVE (Ser307) Polyclonal antibody specifically detects Phospho-PIKFYVE (Ser307) in Human, Mouse samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin).Specifications
Phospho-PIKFYVE (Ser307) | |
Polyclonal | |
Unconjugated | |
PIKFYVE | |
1-phosphatidylinositol 3-phosphate 5-kinase; 1-phosphatidylinositol-3-phosphate 5-kinase; 5230400C17Rik; CFD; epididymis luminal protein 37; FAB1; FYVE finger-containing phosphoinositide kinase; HEL37; Kiaa0981; p235; Phosphatidylinositol 3-phosphate 5-kinase; Phosphatidylinositol 3-phosphate 5-kinase type III; phosphatidylinositol-3-phosphate 5-kinase type III; phosphatidylinositol-3-phosphate/phosphatidylinositol 5-kinase, type III; phosphatidylinositol-4-phosphate 5-kinase, type III; phosphoinositide kinase, FYVE finger containing; phosphoinositide kinase, FYVE-type zinc finger containing; PIKfyve; Pip5k; PIP5K3; Pipk5k3; PipkIII; PIPkin-III; PtdIns(4)P-5-kinase; type III PIP kinase; ZFYVE29; zinc finger, FYVE domain containing 29 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
18711, 200576 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin) | |
1 mg/mL | |
Dulbecco's PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q9Y2I7, Q9Z1T6 | |
PIKFYVE | |
A synthetic phosphopeptide derived from human PIP5K around the phosphorylation site of Ser307 (S-A-SP-I-T) | |
100 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |