missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-HSL (Ser552) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA538254
Code : NEW
Additional Details : Weight : 0.00900kg
Description
In adipose tissue and heart, it primarily hydrolyzes stored triglycerides to free fatty acids, while in steroidogenic tissues, it principally converts cholesteryl esters to free cholesterol for steroid hormone production.Specifications
Phospho-HSL (Ser552) | |
Polyclonal | |
Unconjugated | |
LIPE | |
EC 3.1.1.79, Hormone sensitive lipase, LIPE, LIPS | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
16890, 25330, 3991 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q05469, P54310, P15304 | |
LIPE | |
A synthetic phosphopeptide derived from human HSL around the phosphorylation site of Ser552 (R-R-SP-V-S) | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |