missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NRXN1 Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Brand: Invitrogen PA518730
Code : LD
Additional Details : Weight : 0.01000kg
Description
This antibody is expected to recognize both reported isoforms (NP_004792.1 and NP_620072.1). It is predicted to react with canine, mouse and rat based on sequence homology.
NRXN1 Polyclonal antibody specifically detects NRXN1 in Human samples. It is validated for Flow Cytometry, Western Blot.Specifications
NRXN1 | |
Polyclonal | |
Unconjugated | |
Nrxn1 | |
1700062G21Rik; 9330127H16Rik; A230068P09Rik; alpha-latrotoxin receptor (calcium-dependent); DKFZp313P2036; FLJ35941; Hs.22998; Kiaa0578; mKIAA0578; neurexin 1; Neurexin 1alpha; neurexin I; neurexin I-alpha; Neurexin I-beta; neurexin-1; Neurexin-1-alpha; neurexin-1-beta; non-processed neurexin I-alpha; Nrxn1; Nrxn1b; PTHSL2; SCZD17 | |
Goat | |
Antigen affinity chromatography | |
RUO | |
9378 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Flow Cytometry, Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
P58400, Q9ULB1 | |
Nrxn1 | |
Synthetic peptide sequence (KEKQPSSAKSSNKN) corresponding to the C-terminus amino acids of NRXN1 | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |