missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HLA-ABC Monoclonal Antibody (W6/32), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA170111
This item is not returnable.
View return policy
Description
MA1-70111 detects HLA-ABC in human samples. MA1-70111 has been successfully used in flow cytometry, functional assay, and immunoprecipitation procedures. The MA1-70111 immunogen is purified membrane of human tonsil cells. Store at 4°C short term. For extended storage aliquot and store at -20°C. Avoid Freeze/Thaw Cycles.
HLA-A belongs to the HLA class I heavy chain paralogues. This class I molecule is a heterodimer consisting of a heavy chain and a light chain (beta-2 microglobulin). The heavy chain is anchored in the membrane. Class I molecules play a central role in the immune system by presenting peptides derived from the endoplasmic reticulum lumen. They are expressed in nearly all cells. The heavy chain is approximately 45 kDa and its gene contains 8 exons. Exon 1 encodes the leader peptide, exons 2 and 3 encode the alpha1 and alpha2 domains, which both bind the peptide, exon 4 encodes the alpha3 domain, exon 5 encodes the transmembrane region, and exons 6 and 7 encode the cytoplasmic tail. Polymorphisms within exon 2 and exon 3 are responsible for the peptide binding specificity of each class one molecule. Typing for these polymorphisms is routinely done for bone marrow and kidney transplantation. Hundreds of HLA-A alleles have been described.Specifications
HLA-ABC | |
Monoclonal | |
1 mg/mL | |
HLA-A | |
MHC Class 1, MHC class I, MHC I HLA, HLA-A, HLA-B, HLA-C, MHC HLA-ABC, HLA-B 1502 | |
Mouse | |
Protein G | |
RUO | |
3105, 3106, 3107 | |
4° C | |
Liquid |
Flow Cytometry, Functional Assay, Immunoprecipitation | |
W6/32 | |
Unconjugated | |
P10321, P01889, P05534 | |
HLA-A, HLA-B, HLA-C | |
Purified membrane of human tonsil cells. | |
500 μg | |
Primary | |
Human | |
Antibody | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction