Learn More
MESP1 Monoclonal Antibody (1A3), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA515645
Code : A0
Additional Details : Weight : 0.01000kg
Description
MA5-15645 targets MESP1 in indirect ELISA, WB applications and shows reactivity with Human samples. The MA5-15645 immunogen is purified recombinant fragment of human MESP1 expressed in E. Coli.<br>. MA5-15645 detects MESP1 which has a predicted molecular weight of approximately 28.5kDa.
Mesp1 (mesoderm posterior 1) is a basic helix-loop-helix-type transcription factor, which along with its related proteins, has an essential role in both segmentation and rostro-caudal patterning of somites in the anterior pre-somitic mesoderm. Mesp1 is expressed in many of the precursors of the cardiovascular system and is known to play a role in the process of cardiac morphogenesis.Specifications
MESP1 | |
Monoclonal | |
Conc. Not Determined | |
ascites with 0.03% sodium azide | |
Q9BRJ9 | |
MESP1 | |
Purified recombinant fragment of human MESP1 expressed in E. Coli. | |
RUO | |
55897 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ELISA, Western Blot | |
1A3 | |
Unconjugated | |
MESP1 | |
bHLHc5; Class C basic helix-loop-helix protein 5; mesoderm posterior 1; mesoderm posterior 1 homolog; mesoderm posterior basic helix-loop-helix transcription factor 1; mesoderm posterior bHLH transcription factor 1; mesoderm posterior protein 1; MESP1; MGC10676 | |
Mouse | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |