missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ITGB1BP2 Monoclonal Antibody (308705), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA524025
Code : NEW
Additional Details : Weight : 0.00010kg
Description
Reconstitute at 0.5 mg/mL in sterile PBS.
ITGB1BP2 Monoclonal antibody specifically detects ITGB1BP2 in Human samples. It is validated for Immunohistochemistry, Western Blot.Specifications
ITGB1BP2 | |
Monoclonal | |
0.5 mg/mL | |
PBS with 5% trehalose and No Preservative | |
Q9UKP3 | |
ITGB1BP2 | |
E. coli-derived recombinant human Melusin Ser2-Glu347 | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2b |
Immunohistochemistry, Western Blot | |
308705 | |
Unconjugated | |
ITGB1BP2 | |
Chordc3; integrin beta 1 binding protein (melusin) 2; integrin beta 1 binding protein 2; integrin beta-1-binding protein 2; integrin subunit beta 1 binding protein 2; ITGB1BP; Itgb1bp2; Melusin; MGC119214; MSTP015; RGD1565015 | |
Mouse | |
Protein A/G | |
RUO | |
26548 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |