Learn More
MED6 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA540845
Code : NEW
Additional Details : Weight : 0.00010kg
Description
Peptide sequence: ALLLDLRQKF PPKFVQLKPG EKPVPVDQTK KEAEPIPETV KPEEKETTKN Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 92%; Horse: 100%; Human: 100%; Mouse: 92%; Rabbit: 92%; Rat: 100%
MED6 Polyclonal antibody specifically detects MED6 in Mouse samples. It is validated for ChIP Assay, Western Blot.Specifications
MED6 | |
Polyclonal | |
Unconjugated | |
Med6 | |
1500012F11Rik; Activator-recruited cofactor 33 kDa component; ARC33; AU019660; AV213303; CTD-2540L5.5; hMed6; hypothetical protein LOC505293; MED6; Mediator complex subunit 6; mediator of RNA polymerase II transcriptio subunit 6-like protein; mediator of RNA polymerase II transcription subunit 6; mediator of RNA polymerase II transcription, subunit 6 homolog; NY-REN-28; Renal carcinoma antigen NY-REN-28 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
69792 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
ChIP Assay, Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q921D4 | |
Med6 | |
Synthetic peptide directed towards the middle region of human MED6 | |
100 μL | |
Primary | |
Mouse | |
Antibody | |
IgG |