missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MD-2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA523137
Code : LD
Additional Details : Weight : 0.01000kg
Description
Suggested positive control: human spleen or Ramos.
MD-2 is a gene that encodes a protein involved in the recognition of bacterial lipopolysaccharides (LPS) by the immune system. It is located on chromosome 8 and is part of a larger family of genes that encode for similar proteins. The MD-2 protein forms a complex with Toll-like receptor 4 (TLR4) to recognize and respond to LPS, triggering an immune response. Mutations or variations in the MD-2 gene have been associated with various diseases and conditions, including sepsis and inflammatory disorders.Specifications
MD-2 | |
Polyclonal | |
Unconjugated | |
LY96 | |
Esop1; ESOP-1; LPS co-receptor MD-2; LY96; ly-96; Lymphocyte antigen 96; MD2; MD-2; myeloid differentiation factor-2; myeloid differentiation protein-2; protein MD-2 | |
Rabbit | |
Protein G | |
RUO | |
17087, 23643, 448830 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.5 mg/mL | |
PBS with 0.05% sodium azide | |
Q9JHF9, Q9Y6Y9 | |
LY96 | |
full length protein MD-2 | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |